Product Details

SNP ID
rs559676
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:31910193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAACCTGTAACTGCATATTACC[A/C]ATGCAACGCATGAGCTTTCTTTTTC
Phenotype
MIM: 601584
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTP4A2 PubMed Links

Gene Details

Gene
PTP4A2
Gene Name
protein tyrosine phosphatase type IVA, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195100.1 Intron NP_001182029.1
NM_001195101.1 Intron NP_001182030.1
NM_080391.3 Intron NP_536316.1
XM_005271231.3 Intron XP_005271288.1
XM_005271232.3 Intron XP_005271289.1
XM_017002401.1 Intron XP_016857890.1

View Full Product Details