Product Details

SNP ID
rs670838
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:112026077 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACAGGAACTCCCTTGGCATCCCTAG[C/T]TCCCAATGTGCAATGGAACTGTATA
Phenotype
MIM: 610493 MIM: 608770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DIXDC1 PubMed Links

Gene Details

Gene
DIXDC1
Gene Name
DIX domain containing 1
There are no transcripts associated with this gene.

Gene
DLAT
Gene Name
dihydrolipoamide S-acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001931.4 Intron NP_001922.2
XM_011542647.2 Intron XP_011540949.1

View Full Product Details