Product Details
- SNP ID
-
rs730380
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:1759944 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGAAGAAACAGGCAGAAGAGAAC[A/T]CCTCCCATCCCTCCGCTAATGGCTT
- Phenotype
-
MIM: 607837
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CLN8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111944156] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLN8
- Gene Name
- ceroid-lipofuscinosis, neuronal 8
- Gene
- LOC101927752
- Gene Name
- uncharacterized LOC101927752
There are no transcripts associated with this gene.
View Full Product Details