Product Details
- SNP ID
-
rs541857
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:120214761 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTGACGCAGCAAAGCAGTCACAGT[C/T]GTCAACCCACTGGAGGACACATAGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OAF
PubMed Links
Gene Details
- Gene
- OAF
- Gene Name
- out at first homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178507.2 |
|
Intron |
|
|
NP_848602.1 |
View Full Product Details