Product Details

SNP ID
rs752211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:49495283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTCTGGTACAGCCTGTTTTGGG[A/G]GCCTCTGCCTGGCACACGGGGCCAG
Phenotype
MIM: 604557
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF423 PubMed Links

Gene Details

Gene
ZNF423
Gene Name
zinc finger protein 423
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271620.2 Intron NP_001258549.1
NM_015069.4 Intron NP_055884.2
XM_005255856.4 Intron XP_005255913.1
XM_005255857.4 Intron XP_005255914.1
XM_006721171.3 Intron XP_006721234.1
XM_017023076.1 Intron XP_016878565.1
XM_017023077.1 Intron XP_016878566.1
XM_017023078.1 Intron XP_016878567.1

View Full Product Details