Product Details

SNP ID
rs319602
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:134886474 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGTTGGAGGGCAGGAGGCATTGCTA[C/T]GGAGGTGAGTGGAGTTGGGTAAATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXNDC15 PubMed Links

Gene Details

Gene
TXNDC15
Gene Name
thioredoxin domain containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024715.3 Intron NP_078991.3

View Full Product Details