Product Details

SNP ID
rs498694
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:170147804 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCTTTGAAGGCAGCTGAAGATGA[A/G]TTAAAAAGTTGAATACAATAGTCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL11B PubMed Links

Gene Details

Gene
METTL11B
Gene Name
methyltransferase like 11B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136107.1 Intron NP_001129579.1
XM_011509232.2 Intron XP_011507534.1
XM_011509233.2 Intron XP_011507535.1
XM_011509234.2 Intron XP_011507536.1
Gene
MIR3119-1
Gene Name
microRNA 3119-1
There are no transcripts associated with this gene.

Gene
MIR3119-2
Gene Name
microRNA 3119-2
There are no transcripts associated with this gene.

View Full Product Details