Product Details
- SNP ID
-
rs728375
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
23
- Location
-
Chr.1:48106192 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCCATGCCTCTAATGAAATTAAGC[A/G]TGCGTTATAAAGAAAGATGCACTAT
- Phenotype
-
MIM: 616392
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SKINT1L
PubMed Links
Gene Details
- Gene
- SKINT1L
- Gene Name
- Skint1 like (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details