Product Details

SNP ID
rs164125
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:162396331 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAAATTCAGCAAATCAATTTCA[A/G]TAAACCTTAAAAACCAATTCAACAA
Phenotype
MIM: 608510
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf226 PubMed Links

Gene Details

Gene
C1orf226
Gene Name
chromosome 1 open reading frame 226
There are no transcripts associated with this gene.

Gene
SH2D1B
Gene Name
SH2 domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053282.4 1470 UTR 3 NP_444512.2

View Full Product Details