Product Details

SNP ID
rs341814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:49066244 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGTAGATACTGAAAGACAGCAG[C/T]TTTTGCTACAAGGATACATATTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8orf22 PubMed Links

Gene Details

Gene
C8orf22
Gene Name
chromosome 8 open reading frame 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007176.4 Intron NP_001007177.1
NM_001256596.1 Intron NP_001243525.1
NM_001256597.1 Intron NP_001243526.1
NM_001256598.1 Intron NP_001243527.1

View Full Product Details