Product Details

SNP ID
hCV764449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105970258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCGCACTTCTGAGGGTGGCAGTT[A/G]TAGCCCAGTGAAGTGATAGTCACAT
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM6 PubMed Links

Gene Details

Gene
ADAM6
Gene Name
ADAM metallopeptidase domain 6 (pseudogene)
There are no transcripts associated with this gene.

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

View Full Product Details