Product Details
- SNP ID
-
rs339326
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:116885232 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCCAGCTAGTGATTTATTGTAAG[A/G]TTATTCAGCTTTTCACCAGTAAAGT
- Phenotype
-
MIM: 612659
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RFX6
PubMed Links
Gene Details
- Gene
- RFX6
- Gene Name
- regulatory factor X6
View Full Product Details