Product Details

SNP ID
rs3853680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50569834 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTTGATTCCATGAGGCCGTCAAT[A/G]CGCTGCTCAGCTTCCTAGCTTCCTA
Phenotype
MIM: 176949
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPK4 PubMed Links

Gene Details

Gene
MAPK4
Gene Name
mitogen-activated protein kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292039.1 Intron NP_001278968.1
NM_001292040.1 Intron NP_001278969.1
NM_002747.3 Intron NP_002738.2
XM_005258299.3 Intron XP_005258356.1
XM_011526074.2 Intron XP_011524376.1
XM_011526075.2 Intron XP_011524377.1
XM_011526076.2 Intron XP_011524378.1
XM_011526077.2 Intron XP_011524379.1
XM_011526078.2 Intron XP_011524380.1
XM_017025839.1 Intron XP_016881328.1

View Full Product Details