Product Details

SNP ID
rs657408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130112197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGCGCCGGCCCCTGAGACCTC[A/C]TGGATACTCAGAATCAAGTCCCCAA
Phenotype
MIM: 300775 MIM: 300333
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ELF4 PubMed Links

Gene Details

Gene
ELF4
Gene Name
E74 like ETS transcription factor 4
There are no transcripts associated with this gene.

Gene
RAB33A
Gene Name
RAB33A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004794.2 Intron NP_004785.1
XM_017029963.1 Intron XP_016885452.1

View Full Product Details