Product Details

SNP ID
rs752758
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:113360207 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGACCTTGTATTCCCAACTGTGAT[A/T]AAGCAGAGTATTCTAGTTGGGCTCT
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
BSPRY PubMed Links
Additional Information
For this assay, SNP(s) [rs79435708] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
BSPRY
Gene Name
B-box and SPRY domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317943.1 Intron NP_001304872.1
NM_001317944.1 Intron NP_001304873.1
NM_017688.2 Intron NP_060158.2
XM_006717149.3 Intron XP_006717212.1

View Full Product Details