Product Details

SNP ID
rs496485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12447304 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAACTGAACAAGAAACCAGGACA[A/C]GGAAGGAAGAACTGGAAGCTATGCC
Phenotype
MIM: 609216
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105371998 PubMed Links

Gene Details

Gene
LOC105371998
Gene Name
uncharacterized LOC105371998
There are no transcripts associated with this gene.

Gene
SPIRE1
Gene Name
spire type actin nucleation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128626.1 5015 UTR 3 NP_001122098.1
NM_001128627.1 5015 UTR 3 NP_001122099.1
NM_020148.2 5015 UTR 3 NP_064533.3
XM_005258122.4 5015 UTR 3 XP_005258179.1
XM_011525701.1 5015 UTR 3 XP_011524003.1
XM_011525702.1 5015 UTR 3 XP_011524004.1
XM_011525703.2 5015 Intron XP_011524005.1

View Full Product Details