Product Details

SNP ID
rs238113
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:3498068 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAAGGAGAAAATATATCTGAAATA[C/T]GAACTACTTGGGTTAAAATAGACGG
Phenotype
MIM: 605445
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DLGAP1 PubMed Links

Gene Details

Gene
DLGAP1
Gene Name
DLG associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003809.2 3278 UTR 3 NP_001003809.1
NM_001242761.1 3278 Intron NP_001229690.1
NM_001242762.1 3278 UTR 3 NP_001229691.1
NM_001242763.1 3278 UTR 3 NP_001229692.1
NM_001242764.1 3278 UTR 3 NP_001229693.1
NM_001242765.1 3278 Intron NP_001229694.1
NM_001242766.1 3278 UTR 3 NP_001229695.1
NM_001308390.1 3278 UTR 3 NP_001295319.1
NM_004746.3 3278 UTR 3 NP_004737.2
XM_005258171.2 3278 UTR 3 XP_005258228.1
XM_005258172.1 3278 UTR 3 XP_005258229.1
XM_005258173.3 3278 UTR 3 XP_005258230.2
XM_005258174.4 3278 UTR 3 XP_005258231.1
XM_006722367.3 3278 UTR 3 XP_006722430.1
XM_011525770.2 3278 Intron XP_011524072.1
XM_011525771.2 3278 Intron XP_011524073.1
XM_017026080.1 3278 UTR 3 XP_016881569.1
XM_017026081.1 3278 UTR 3 XP_016881570.1
XM_017026082.1 3278 Intron XP_016881571.1
XM_017026083.1 3278 UTR 3 XP_016881572.1
XM_017026084.1 3278 Intron XP_016881573.1
XM_017026085.1 3278 Intron XP_016881574.1

View Full Product Details