Product Details

SNP ID
rs532242
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:52842993 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTACAGGTGAGCACCGGGGTGCGGG[C/G]GGCGACGCGGACCTCGGCGCCACGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZYG11A PubMed Links

Gene Details

Gene
ZYG11A
Gene Name
zyg-11 family member A, cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004339.2 Intron NP_001004339.2
NM_001307931.1 Intron NP_001294860.1
XM_011541480.2 Intron XP_011539782.1
XM_011541481.2 Intron XP_011539783.1
XM_011541482.2 Intron XP_011539784.1
XM_011541483.2 Intron XP_011539785.1
XM_011541484.1 Intron XP_011539786.1
XM_011541485.1 Intron XP_011539787.1
XM_011541486.1 Intron XP_011539788.1
XM_011541487.2 Intron XP_011539789.1
XM_011541488.1 Intron XP_011539790.1

View Full Product Details