Product Details

SNP ID
rs694213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40896089 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGATACACTGCTCCGGTAATCAAG[A/G]GCTCACAGAGCTTAGGAGTAGGGAC
Phenotype
MIM: 608551
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS18 PubMed Links

Gene Details

Gene
VPS18
Gene Name
VPS18, CORVET/HOPS core subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020857.2 601 Intron NP_065908.1
XM_011521843.2 601 Intron XP_011520145.1
XM_011521844.2 601 UTR 5 XP_011520146.1
XM_017022447.1 601 Intron XP_016877936.1

View Full Product Details