Product Details

SNP ID
rs121909555
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:173903968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTCTCTTATAAAAACCAGGAAA[G/A]GCCTGTTGGCCTTGAAAGTCACCCT
Phenotype
MIM: 107300
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
SERPINC1 PubMed Links

Gene Details

Gene
SERPINC1
Gene Name
serpin family C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000488.3 1435 Missense Mutation CCT,CTT P439L NP_000479.1
XM_005245198.3 1435 Missense Mutation CCT,CTT P391L XP_005245255.1

View Full Product Details