Product Details
- SNP ID
-
rs283508
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:49972860 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCACACGGGCCAGTGTTGTCAGTC[C/T]CCAACTCTGGACCAATGTCCAGGCT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SIGLEC16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs192711450] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SIGLEC16
- Gene Name
- sialic acid binding Ig like lectin 16 (gene/pseudogene)
There are no transcripts associated with this gene.
- Gene
- VRK3
- Gene Name
- vaccinia related kinase 3
There are no transcripts associated with this gene.
View Full Product Details