Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_173563.2 | Intron | NP_775834.2 | |||
XM_005248951.3 | Intron | XP_005249008.1 | |||
XM_006715025.2 | Intron | XP_006715088.1 | |||
XM_006715026.3 | Intron | XP_006715089.1 | |||
XM_011514413.2 | Intron | XP_011512715.1 | |||
XM_011514414.2 | Intron | XP_011512716.1 | |||
XM_011514415.1 | Intron | XP_011512717.1 | |||
XM_011514416.2 | Intron | XP_011512718.1 | |||
XM_011514417.1 | Intron | XP_011512719.1 | |||
XM_017010480.1 | Intron | XP_016865969.1 | |||
XM_017010481.1 | Intron | XP_016865970.1 | |||
XM_017010482.1 | Intron | XP_016865971.1 |