Product Details

SNP ID
rs638845
Assay Type
Functionally Tested
NCBI dbSNP Submissions
40
Location
Chr.1:46674704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGATTCTGTGTTGCGGGGAGGGG[A/G]GGAGGTATCCAGAATCTAGCAAACA
Phenotype
MIM: 608917
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ATPAF1 PubMed Links
Additional Information
For this assay, SNP(s) [rs76149447] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ATPAF1
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
There are no transcripts associated with this gene.

Gene
EFCAB14
Gene Name
EF-hand calcium binding domain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014774.2 Intron NP_055589.1
Gene
EFCAB14-AS1
Gene Name
EFCAB14 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TEX38
Gene Name
testis expressed 38
There are no transcripts associated with this gene.

View Full Product Details