Product Details

SNP ID
rs269298
Assay Type
Functionally Tested
NCBI dbSNP Submissions
42
Location
Chr.1:52726515 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCTGCGGCTGCTACTGCTACGCT[C/T]CTAGCTTGAGGGAAAGAGGCCGAGG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZYG11B PubMed Links

Gene Details

Gene
ZYG11B
Gene Name
zyg-11 family member B, cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024646.2 62 UTR 5 NP_078922.1
XM_006710898.3 62 Intron XP_006710961.1
XM_017002336.1 62 UTR 5 XP_016857825.1

View Full Product Details