Product Details

SNP ID
rs483281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31820831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTCCCCTCCCGGCCTCTGTCG[C/T]TTTTCCTCTCTCCCCTTAACTACTT
Phenotype
MIM: 607108
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DKFZp686K1684 PubMed Links

Gene Details

Gene
DKFZp686K1684
Gene Name
uncharacterized LOC440034
There are no transcripts associated with this gene.

Gene
PAUPAR
Gene Name
PAX6 upstream antisense RNA
There are no transcripts associated with this gene.

Gene
PAX6
Gene Name
paired box 6
There are no transcripts associated with this gene.

View Full Product Details