Product Details

SNP ID
rs13237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9969747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGCTTTGTTTGTGTCTGCGGGC[G/A]TGGCCATGGCTGCTCCCGTCATCTG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
EMC3 PubMed Links

Gene Details

Gene
EMC3
Gene Name
ER membrane protein complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018447.3 1096 Missense Mutation ACG,ATG T210M NP_060917.1
XM_005265321.3 1096 Missense Mutation ACG,ATG T210M XP_005265378.1

View Full Product Details