Product Details

SNP ID
rs746693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:7583374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTGAGGCCTCAACACATTTCAG[A/G]ATGTCCCATGGACAGTGGCTACAGC
Phenotype
MIM: 176888
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPRM PubMed Links

Gene Details

Gene
PTPRM
Gene Name
protein tyrosine phosphatase, receptor type M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105244.1 Intron NP_001098714.1
NM_002845.3 Intron NP_002836.3
XM_011525708.2 Intron XP_011524010.1
XM_011525710.2 Intron XP_011524012.1
XM_011525712.2 Intron XP_011524014.1
XM_011525713.2 Intron XP_011524015.1
XM_011525715.2 Intron XP_011524017.1
XM_011525716.2 Intron XP_011524018.1
XM_011525717.2 Intron XP_011524019.1
XM_011525722.2 Intron XP_011524024.1
XM_017025894.1 Intron XP_016881383.1
XM_017025895.1 Intron XP_016881384.1
XM_017025896.1 Intron XP_016881385.1
XM_017025897.1 Intron XP_016881386.1
XM_017025898.1 Intron XP_016881387.1
XM_017025899.1 Intron XP_016881388.1
XM_017025900.1 Intron XP_016881389.1
XM_017025901.1 Intron XP_016881390.1
XM_017025902.1 Intron XP_016881391.1
XM_017025903.1 Intron XP_016881392.1
XM_017025904.1 Intron XP_016881393.1
XM_017025905.1 Intron XP_016881394.1
XM_017025906.1 Intron XP_016881395.1
XM_017025907.1 Intron XP_016881396.1
XM_017025908.1 Intron XP_016881397.1
XM_017025909.1 Intron XP_016881398.1
XM_017025910.1 Intron XP_016881399.1
XM_017025911.1 Intron XP_016881400.1
XM_017025912.1 Intron XP_016881401.1

View Full Product Details