Product Details

SNP ID
rs203463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19908386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTACCCAATGCCTTGTGTATTAC[A/T]AAGTCTCTCTACTCTGGCCAGATCC
Phenotype
MIM: 604694
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AKAP10 PubMed Links

Gene Details

Gene
AKAP10
Gene Name
A-kinase anchoring protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007202.3 Intron NP_009133.2
XM_006721432.3 Intron XP_006721495.1
XM_017024071.1 Intron XP_016879560.1

View Full Product Details