Product Details

SNP ID
rs297118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55773622 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTCAAAACTCCATACAATCATGT[A/G]CTTTTTCCTTAGTCACTTGTCCTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5D13 PubMed Links

Gene Details

Gene
OR5D13
Gene Name
olfactory receptor family 5 subfamily D member 13 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001967.1 185 Missense Mutation TAC,TGC Y62C NP_001001967.1

View Full Product Details