Product Details
- SNP ID
-
rs376240
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35870188 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGGCGTGGCCATGAAAAGACAAAT[C/T]TATAACGGGAAGGGAGAGTTTTGGA
- Phenotype
-
MIM: 104775
MIM: 607762
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APLP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115712630] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APLP1
- Gene Name
- amyloid beta precursor like protein 1
- Gene
- KIRREL2
- Gene Name
- kin of IRRE like 2 (Drosophila)
There are no transcripts associated with this gene.
View Full Product Details