Product Details

SNP ID
rs271193
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:12131817 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACAGAAGTATTTGTTGCCAAGAAAC[C/T]GCATTAGTGTTTTTAGTCAGGTATA
Phenotype
MIM: 194557
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF20 PubMed Links

Gene Details

Gene
ZNF20
Gene Name
zinc finger protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203250.1 2594 UTR 3 NP_001190179.1
NM_021143.3 2594 UTR 3 NP_066966.2
Gene
ZNF625-ZNF20
Gene Name
ZNF625-ZNF20 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details