Product Details

SNP ID
rs360615
Assay Type
Validated
NCBI dbSNP Submissions
57
Location
Chr.1:114858515 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTAGTTTGGTTATATTAGAATTTTG[A/G]ACAATTAATTTTTGAAAACATATTT
Phenotype
MIM: 602162
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYCP1 PubMed Links

Gene Details

Gene
SYCP1
Gene Name
synaptonemal complex protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282541.1 Intron NP_001269470.1
NM_001282542.1 Intron NP_001269471.1
NM_003176.3 Intron NP_003167.2
XM_005271155.3 Intron XP_005271212.1
XM_005271156.1 Intron XP_005271213.1
XM_006710859.1 Intron XP_006710922.1
XM_011542037.2 Intron XP_011540339.1
XM_011542038.2 Intron XP_011540340.1
XM_011542039.1 Intron XP_011540341.1
XM_017002184.1 Intron XP_016857673.1
XM_017002185.1 Intron XP_016857674.1

View Full Product Details