Product Details
- SNP ID
-
rs637990
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
62
- Location
-
Chr.1:20721123 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- GATTCTTGGCCCTGTCTGCTGGTGC[A/G]CATCCCTTTCTGCACACAGGAGTCC
- Phenotype
-
MIM: 605037
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KIF17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75869282] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KIF17
- Gene Name
- kinesin family member 17
There are no transcripts associated with this gene.
- Gene
- SH2D5
- Gene Name
- SH2 domain containing 5
View Full Product Details