Product Details
- SNP ID
-
rs60986317
- Assay Type
- DME
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:51934853 on Build GRCh38
- Set Membership
-
DME
Validated
Inventoried
- Context Sequence [VIC/FAM]
- GCTGTGCCGAGATGGCTTGTCGGAC[A/G]TCAGGGAGGACAGCGACACCTGGCT
- Phenotype
-
MIM: 606882
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATP7B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116091486] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP7B
- Gene Name
- ATPase copper transporting beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000053.3 |
3837 |
Missense Mutation |
ACG,ATG |
T1434M |
NP_000044.2 |
NM_001005918.2 |
3837 |
Missense Mutation |
ACG,ATG |
T1227M |
NP_001005918.1 |
NM_001243182.1 |
3837 |
Missense Mutation |
ACG,ATG |
T1323M |
NP_001230111.1 |
XM_005266423.2 |
3837 |
Missense Mutation |
ACG,ATG |
T1402M |
XP_005266480.1 |
XM_005266424.4 |
3837 |
Missense Mutation |
ACG,ATG |
T1402M |
XP_005266481.1 |
XM_005266427.2 |
3837 |
Missense Mutation |
ACG,ATG |
T1356M |
XP_005266484.1 |
XM_005266428.1 |
3837 |
Missense Mutation |
ACG,ATG |
T1350M |
XP_005266485.1 |
XM_005266430.4 |
3837 |
Missense Mutation |
ACG,ATG |
T1434M |
XP_005266487.1 |
XM_005266431.3 |
3837 |
Missense Mutation |
ACG,ATG |
T1422M |
XP_005266488.1 |
XM_005266432.2 |
3837 |
Missense Mutation |
ACG,ATG |
T1272M |
XP_005266489.1 |
XM_006719837.3 |
3837 |
Missense Mutation |
ACG,ATG |
T1402M |
XP_006719900.1 |
XM_006719838.1 |
3837 |
Missense Mutation |
ACG,ATG |
T706M |
XP_006719901.1 |
XM_006719839.1 |
3837 |
Missense Mutation |
ACG,ATG |
T645M |
XP_006719902.1 |
XM_011535117.2 |
3837 |
Missense Mutation |
ACG,ATG |
T1402M |
XP_011533419.1 |
XM_011535118.1 |
3837 |
Missense Mutation |
ACG,ATG |
T1389M |
XP_011533420.1 |
XM_011535119.1 |
3837 |
Missense Mutation |
ACG,ATG |
T1373M |
XP_011533421.1 |
XM_011535122.2 |
3837 |
Missense Mutation |
ACG,ATG |
T990M |
XP_011533424.1 |
XM_017020627.1 |
3837 |
Missense Mutation |
ACG,ATG |
T1402M |
XP_016876116.1 |
XM_017020628.1 |
3837 |
Missense Mutation |
ACG,ATG |
T990M |
XP_016876117.1 |
View Full Product Details