Product Details
- SNP ID
-
rs304074
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:4494290 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCTTCGCCCAAGCGATCCCACCAAC[C/T]TGCCTACCAAGTTTGGGGGCGGGGG
- Phenotype
-
MIM: 147265
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ITPR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs184340186] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ITPR1
- Gene Name
- inositol 1,4,5-trisphosphate receptor type 1
- Gene
- ITPR1-AS1
- Gene Name
- ITPR1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details