Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001171740.2 | 2220 | UTR 3 | NP_001165211.1 | ||
NM_001171741.2 | 2220 | UTR 3 | NP_001165212.1 | ||
NM_001171743.2 | 2220 | UTR 3 | NP_001165214.2 | ||
NM_016210.4 | 2220 | UTR 3 | NP_057294.2 | ||
XM_011533781.2 | 2220 | UTR 3 | XP_011532083.1 | ||
XM_011533782.2 | 2220 | UTR 3 | XP_011532084.1 | ||
XM_011533783.2 | 2220 | UTR 3 | XP_011532085.1 | ||
XM_011533784.2 | 2220 | UTR 3 | XP_011532086.1 | ||
XM_011533785.2 | 2220 | UTR 3 | XP_011532087.1 | ||
XM_011533788.1 | 2220 | UTR 3 | XP_011532090.1 | ||
XM_011533789.1 | 2220 | UTR 3 | XP_011532091.1 | ||
XM_011533790.1 | 2220 | UTR 3 | XP_011532092.1 | ||
XM_017006545.1 | 2220 | UTR 3 | XP_016862034.1 | ||
XM_017006546.1 | 2220 | UTR 3 | XP_016862035.1 | ||
XM_017006547.1 | 2220 | UTR 3 | XP_016862036.1 |