Product Details

SNP ID
rs766783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114398623 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAATAAATATCTCTATAAACAATA[A/G]AAGTCCTAGCACACTGACTTGCACA
Phenotype
MIM: 612413
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf71 PubMed Links

Gene Details

Gene
C11orf71
Gene Name
chromosome 11 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271562.1 1928 UTR 3 NP_001258491.1
NM_019021.3 1928 Intron NP_061894.2
Gene
RBM7
Gene Name
RNA binding motif protein 7
There are no transcripts associated with this gene.

View Full Product Details