Product Details

SNP ID
rs210030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130122430 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTGGCATTGTTTGCCTGTATGCA[A/C]TTTCCTTTTGTCTGTATGATAAGAG
Phenotype
MIM: 300169 MIM: 310490 MIM: 300333
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AIFM1 PubMed Links

Gene Details

Gene
AIFM1
Gene Name
apoptosis inducing factor, mitochondria associated 1
There are no transcripts associated with this gene.

Gene
RAB33A
Gene Name
RAB33A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004794.2 Intron NP_004785.1
XM_017029963.1 Intron XP_016885452.1

View Full Product Details