Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001272003.1 | 133 | Intron | NP_001258932.1 | ||
NM_020822.2 | 133 | Intron | NP_065873.2 | ||
XM_011518877.2 | 133 | UTR 5 | XP_011517179.1 | ||
XM_011518878.2 | 133 | UTR 5 | XP_011517180.1 | ||
XM_011518879.2 | 133 | UTR 5 | XP_011517181.1 | ||
XM_011518880.1 | 133 | Intron | XP_011517182.1 | ||
XM_011518881.2 | 133 | Intron | XP_011517183.1 | ||
XM_017014931.1 | 133 | Intron | XP_016870420.1 | ||
XM_017014932.1 | 133 | Intron | XP_016870421.1 | ||
XM_017014933.1 | 133 | Intron | XP_016870422.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001012415.2 | 133 | Intron | NP_001012415.2 | ||
NM_001101677.1 | 133 | Intron | NP_001095147.1 | ||
XM_005263403.3 | 133 | Intron | XP_005263460.1 | ||
XM_006717109.3 | 133 | UTR 5 | XP_006717172.1 | ||
XM_011518698.2 | 133 | UTR 5 | XP_011517000.1 |