Product Details
- SNP ID
-
hCV13304
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:153182483 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCCTGGTGTGTGTGCAGGCTGCC[G/A]CCTCCTCCTCCTCTCCTCTGGTCCT
- Phenotype
-
MIM: 300016
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAGEA1
PubMed Links
Gene Details
- Gene
- MAGEA1
- Gene Name
- MAGE family member A1
There are no transcripts associated with this gene.
View Full Product Details