Product Details

SNP ID
rs10795207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:4873120 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGAACCCAGTGGGAGGTGGTTGA[A/G]TTATGAGGGTGGGTTTTTCCTGCGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKR1C6P PubMed Links

Gene Details

Gene
AKR1C6P
Gene Name
aldo-keto reductase family 1 member C6, pseudogene
There are no transcripts associated with this gene.

Gene
AKR1E2
Gene Name
aldo-keto reductase family 1 member E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040177.2 Intron NP_001035267.1
NM_001271021.1 Intron NP_001257950.1
NM_001271025.1 Intron NP_001257954.1
XM_011519715.2 Intron XP_011518017.1
XM_011519718.2 Intron XP_011518020.1
XM_011519719.2 Intron XP_011518021.1
XM_011519720.2 Intron XP_011518022.1
XM_011519722.2 Intron XP_011518024.1
XM_011519724.2 Intron XP_011518026.1
XM_011519725.2 Intron XP_011518027.1
XM_011519729.2 Intron XP_011518031.1
XM_017016743.1 Intron XP_016872232.1
XM_017016744.1 Intron XP_016872233.1
XM_017016745.1 Intron XP_016872234.1

View Full Product Details