Product Details

SNP ID
rs12631122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129400675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGTGAGATTTTTGGGTGTGAGG[A/G]GAATGGGGAGTTTGAGGAGACCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFCAB12 PubMed Links

Gene Details

Gene
EFCAB12
Gene Name
EF-hand calcium binding domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207307.2 Intron NP_997190.1
XM_011513293.2 Intron XP_011511595.1
XM_011513294.2 Intron XP_011511596.1
XM_011513295.2 Intron XP_011511597.1
XM_011513297.2 Intron XP_011511599.1
XM_011513298.1 Intron XP_011511600.1
XM_011513299.2 Intron XP_011511601.1
Gene
RPL32P3
Gene Name
ribosomal protein L32 pseudogene 3
There are no transcripts associated with this gene.

Gene
SNORA7B
Gene Name
small nucleolar RNA, H/ACA box 7B
There are no transcripts associated with this gene.

View Full Product Details