Product Details

SNP ID
rs4796856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81941396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCACAGCCGGCGGGCTCGGGGG[G/A]ACACTCCCGCCGGGCAAAGTACAGC
Phenotype
MIM: 179035
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MYADML2 PubMed Links

Gene Details

Gene
MYADML2
Gene Name
myeloid associated differentiation marker like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145113.2 701 Missense Mutation CCC,TCC P116S NP_001138585.2
Gene
PYCR1
Gene Name
pyrroline-5-carboxylate reductase 1
There are no transcripts associated with this gene.

View Full Product Details