Product Details

SNP ID
rs3210749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:158667579 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAAATGAGCAAAAATGCCTGTA[A/C]TCCCAGTGCTTTATAGGAGGAGGCG
Phenotype
MIM: 616210 MIM: 231675
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C4orf46 PubMed Links

Gene Details

Gene
C4orf46
Gene Name
chromosome 4 open reading frame 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008393.3 2625 UTR 3 NP_001008394.1
Gene
ETFDH
Gene Name
electron transfer flavoprotein dehydrogenase
There are no transcripts associated with this gene.

View Full Product Details