Product Details

SNP ID
rs3809357
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:20566642 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTCCAAGGTTCACAAGGCTCGTGC[A/T]GCGGCTCCGGGAGTTATGTCACAGT
Phenotype
MIM: 600595
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IFT88 PubMed Links

Gene Details

Gene
IFT88
Gene Name
intraflagellar transport 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318491.1 197 Intron NP_001305420.1
NM_001318493.1 197 Intron NP_001305422.1
NM_006531.4 197 Intron NP_006522.2
NM_175605.4 197 Intron NP_783195.2
XM_005266553.2 197 UTR 5 XP_005266610.1
XM_006719870.3 197 Intron XP_006719933.1
XM_011535241.2 197 Intron XP_011533543.1
XM_011535242.1 197 UTR 5 XP_011533544.1
XM_011535243.1 197 UTR 5 XP_011533545.1
XM_017020757.1 197 UTR 5 XP_016876246.1
XM_017020758.1 197 Intron XP_016876247.1
XM_017020759.1 197 Intron XP_016876248.1
XM_017020760.1 197 Intron XP_016876249.1
XM_017020761.1 197 Intron XP_016876250.1
XM_017020762.1 197 Intron XP_016876251.1
XM_017020763.1 197 Intron XP_016876252.1
XM_017020764.1 197 Intron XP_016876253.1
XM_017020765.1 197 Intron XP_016876254.1
XM_017020766.1 197 Intron XP_016876255.1
XM_017020767.1 197 Intron XP_016876256.1
XM_017020768.1 197 Intron XP_016876257.1
XM_017020769.1 197 Intron XP_016876258.1
XM_017020770.1 197 Intron XP_016876259.1
XM_017020771.1 197 UTR 5 XP_016876260.1
XM_017020772.1 197 Intron XP_016876261.1
XM_017020773.1 197 UTR 5 XP_016876262.1
XM_017020774.1 197 Intron XP_016876263.1
XM_017020775.1 197 Intron XP_016876264.1
XM_017020776.1 197 Intron XP_016876265.1

View Full Product Details