Product Details

SNP ID
rs7626551
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:52904076 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TACCCTCCAAAACATGTTTTCTGTG[C/G]TGTGCTTCTACACTGGAGACACCTG
Phenotype
MIM: 607319
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SFMBT1 PubMed Links

Gene Details

Gene
SFMBT1
Gene Name
Scm-like with four mbt domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016329.3 4287 UTR 3 NP_057413.2
XM_005265221.3 4287 UTR 3 XP_005265278.1
XM_006713203.3 4287 UTR 3 XP_006713266.1
XM_006713204.3 4287 UTR 3 XP_006713267.1
XM_011533824.2 4287 UTR 3 XP_011532126.1
XM_011533825.2 4287 UTR 3 XP_011532127.1
Gene
TMEM110
Gene Name
transmembrane protein 110
There are no transcripts associated with this gene.

Gene
TMEM110-MUSTN1
Gene Name
TMEM110-MUSTN1 readthrough
There are no transcripts associated with this gene.

View Full Product Details