Product Details

SNP ID
rs13150454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109835055 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTGTCTTTTCCTCCTGTTGAATT[A/G]CCCATCTTCTGAATACTAGTCTTTT
Phenotype
MIM: 605224
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RRH PubMed Links

Gene Details

Gene
RRH
Gene Name
retinal pigment epithelium-derived rhodopsin homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006583.2 Intron NP_006574.1

View Full Product Details