Product Details
- SNP ID
-
rs11587728
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:16922992 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGTAATTTGTCCAGACTTATTCC[G/T]CATGTGGTGCAACTAGAAGGAAGGG
- Phenotype
-
MIM: 615776
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CROCC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11576698] are located under a probe and SNP(s) [rs377726134] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CROCC
- Gene Name
- ciliary rootlet coiled-coil, rootletin
View Full Product Details