Product Details

SNP ID
rs12570598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5065603 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCGAAATGAAAGTACACTTCACAG[G/T]GTGGTAATGGGCATGAGCATAGGGC
Phenotype
MIM: 603966
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AKR1C3 PubMed Links

Gene Details

Gene
AKR1C3
Gene Name
aldo-keto reductase family 1, member C3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253908.1 Intron NP_001240837.1
NM_001253909.1 Intron NP_001240838.1
NM_003739.5 Intron NP_003730.4

View Full Product Details