Product Details

SNP ID
rs6427671
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:153664928 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCACTTGTCTAAAAATCACTAGAA[A/G]CATGCAAGATCTGTTTTCTTCAAAA
Phenotype
MIM: 603181 MIM: 607007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ILF2 PubMed Links

Gene Details

Gene
ILF2
Gene Name
interleukin enhancer binding factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267809.1 Intron NP_001254738.1
NM_004515.3 Intron NP_004506.2
Gene
SNAPIN
Gene Name
SNAP associated protein
There are no transcripts associated with this gene.

View Full Product Details